Methyl 2'-fluoro-1,1'-biphenyl-4-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
PC300619-1G
PC300619-5G
APOSPC300619-1GEA
248
EUR
APOSPC300619-1G
APOSPC300619-5G
Methyl 2'-fluoro-1,1'-biphenyl-4-carboxylate
Methyl 2'-fluoro-1,1'-biphenyl-4-carboxylate
Formel:
C₁₄H₁₁FO₂ Molecular Weight: 230,234 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00662461 CAS-Nummer: 80254-85-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...