1-[4-(Trifluoromethyl)pyridin-2-yl]piperazine

Lieferant: Apollo Scientific
Achtung

PC300626-1G PC300626-500MG PC300626-5G
APOSPC300626-1GEA 314 EUR
APOSPC300626-1G APOSPC300626-500MG APOSPC300626-5G
1-[4-(Trifluoromethyl)pyridin-2-yl]piperazine
1-[4-(Trifluoromethyl)pyridin-2-yl]piperazine
Formel: C₁₀H₁₂F₃N₃
Molecular Weight: 231,218 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00114707
CAS-Nummer: 118708-88-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR