1-(3-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}phenyl)ethanone
Lieferant: Apollo Scientific
PC300646-1G
PC300646-10G
PC300646-5G
APOSPC300646-1GEA
193
EUR
APOSPC300646-1G
APOSPC300646-10G
APOSPC300646-5G
1-(3-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}phenyl)ethanone
1-(3-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}phenyl)ethanone
Formel:
C₁₃H₉F₃N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11846259 CAS-Nummer: 1227954-39-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...