Ethyl 4-{[4-(trifluoromethyl)pyrimidin-2-yl]oxy}benzoate

Lieferant: Apollo Scientific

PC300678-1G PC300678-5G PC300678-10G
APOSPC300678-1GEA 161 EUR
APOSPC300678-1G APOSPC300678-5G APOSPC300678-10G
Ethyl 4-{[4-(trifluoromethyl)pyrimidin-2-yl]oxy}benzoate
Ethyl 4-{[4-(trifluoromethyl)pyrimidin-2-yl]oxy}benzoate
Formel: C₁₄H₁₁F₃N₂O₃
Molecular Weight: 312.244 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14583151
CAS-Nummer: 1259324-17-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR