1,1,1-Trifluoro-4-(4-phenylpiperazin-1-yl)but-3-en-2-one

Lieferant: Apollo Scientific

PC300679-1G PC300679-5G PC300679-10G
APOSPC300679-1GEA 161 EUR
APOSPC300679-1G APOSPC300679-5G APOSPC300679-10G
1,1,1-Trifluoro-4-(4-phenylpiperazin-1-yl)but-3-en-2-one
1,1,1-Trifluoro-4-(4-phenylpiperazin-1-yl)but-3-en-2-one
Formel: C₁₄H₁₅F₃N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD02186476
CAS-Nummer: 478047-04-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR