2-[(2,4-Difluorophenoxy)methyl]oxirane

Lieferant: Apollo Scientific

PC300681-1G PC300681-5G PC300681-10G
APOSPC300681-1GEA 173 EUR
APOSPC300681-1G APOSPC300681-5G APOSPC300681-10G
2-[(2,4-Difluorophenoxy)methyl]oxirane
2-[(2,4-Difluorophenoxy)methyl]oxirane
Formel: C₉H₈F₂O₂
Molecular Weight: 186,155 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09741082
CAS-Nummer: 169286-61-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR