2-{4-[4-(Trifluoromethyl)pyrimidin-2-yl]piperazin-1-yl}ethanol

Lieferant: Apollo Scientific

PC300683-1G PC300683-5G PC300683-10G
APOSPC300683-1GEA 147 EUR
APOSPC300683-1G APOSPC300683-5G APOSPC300683-10G
2-{4-[4-(Trifluoromethyl)pyrimidin-2-yl]piperazin-1-yl}ethanol
2-{4-[4-(Trifluoromethyl)pyrimidin-2-yl]piperazin-1-yl}ethanol
Formel: C₁₁H₁₅F₃N₄O
Molecular Weight: 276.258 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01312087
CAS-Nummer: 651004-99-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR