3-Bromo-4-(trifluoromethyl)-1H-indazole

Lieferant: Apollo Scientific
Achtung

PC300706-1G PC300706-250MG
APOSPC300706-1GEA 290 EUR
APOSPC300706-1G APOSPC300706-250MG
3-Bromo-4-(trifluoromethyl)-1H-indazole
3-Bromo-4-(trifluoromethyl)-1H-indazole
Formel: C₈H₄BrF₃N₂
Molecular Weight: 265.03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09749936
CAS-Nummer: 1000342-21-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR