Ethyl 2-nitro-4-(trifluoromethyl)benzoate

Lieferant: Apollo Scientific

PC300712-1G PC300712-5G
APOSPC300712-1GEA 102 EUR
APOSPC300712-1G APOSPC300712-5G
Ethyl 2-nitro-4-(trifluoromethyl)benzoate
Ethyl 2-nitro-4-(trifluoromethyl)benzoate
Formel: C₁₀H₈F₃NO₄
Molecular Weight: 263,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12406841
CAS-Nummer: 702673-02-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR