Methyl 2,3,5-trifluoro-4-pyrrolidin-1-ylbenzoate

Lieferant: Apollo Scientific

PC300763-1G PC300763-5G PC300763-250MG
APOSPC300763-1GEA 232 EUR
APOSPC300763-1G APOSPC300763-5G APOSPC300763-250MG
Methyl 2,3,5-trifluoro-4-pyrrolidin-1-ylbenzoate
Methyl 2,3,5-trifluoro-4-pyrrolidin-1-ylbenzoate
Formel: C₁₂H₁₂F₃NO₂
Molecular Weight: 259,224 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28040514
CAS-Nummer: 1858257-29-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR