N-(2-Chloro-6-fluorobenzyl)-N-methylamine

Lieferant: Apollo Scientific

PC300798-1G PC300798-250MG
APOSPC300798-1GEA 174 EUR
APOSPC300798-1G APOSPC300798-250MG
N-(2-Chloro-6-fluorobenzyl)-N-methylamine
N-(2-Chloro-6-fluorobenzyl)-N-methylamine
Formel: C₈H₉ClFN
Molecular Weight: 173,615 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00013628
CAS-Nummer: 62924-64-5
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR