3-Amino-3-[4-(trifluoromethoxy)phenyl]propanoic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC300802-1G
PC300802-5G
APOSPC300802-1GEA
386
EUR
APOSPC300802-1G
APOSPC300802-5G
3-Amino-3-[4-(trifluoromethoxy)phenyl]propanoic acid
3-Amino-3-[4-(trifluoromethoxy)phenyl]propanoic acid
Formel:
C₁₀H₁₀F₃NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00764454 CAS-Nummer: 213192-56-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...