N-[4-(3-Fluorophenoxy)benzyl]-N-methylamine

Lieferant: Apollo Scientific

PC300812-1G PC300812-5G
APOSPC300812-1GEA 296 EUR
APOSPC300812-1G APOSPC300812-5G
N-[4-(3-Fluorophenoxy)benzyl]-N-methylamine
N-[4-(3-Fluorophenoxy)benzyl]-N-methylamine
Formel: C₁₄H₁₄FNO
Molecular Weight: 231.265 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11631331
CAS-Nummer: 1095078-57-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR