Methyl 2-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzoate

Lieferant: Apollo Scientific

PC300862-1G PC300862-5G
APOSPC300862-1GEA 186 EUR
APOSPC300862-1G APOSPC300862-5G
Methyl 2-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzoate
Methyl 2-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzoate
Formel: C₁₄H₉ClF₃NO₃
Molecular Weight: 331,674 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03644664
CAS-Nummer: 852952-21-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR