1,1,1-Trifluoro-3-pyrrolidin-1-ylpropan-2-ol

Lieferant: Apollo Scientific
Achtung

PC300873-1G PC300873-250MG PC300873-5G
APOSPC300873-1GEA 58 EUR
APOSPC300873-1G APOSPC300873-250MG APOSPC300873-5G
1,1,1-Trifluoro-3-pyrrolidin-1-ylpropan-2-ol
1,1,1-Trifluoro-3-pyrrolidin-1-ylpropan-2-ol
Formel: C₇H₁₂F₃NO
Molecular Weight: 183,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00466829
CAS-Nummer: 959045-77-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR