1,1,1-Trifluoro-3-(4-methylpiperazin-1-yl)propan-2-ol

Lieferant: Apollo Scientific
Achtung

PC300874-1G PC300874-250MG PC300874-5G
APOSPC300874-1GEA 265 EUR
APOSPC300874-1G APOSPC300874-250MG APOSPC300874-5G
1,1,1-Trifluoro-3-(4-methylpiperazin-1-yl)propan-2-ol
1,1,1-Trifluoro-3-(4-methylpiperazin-1-yl)propan-2-ol
Formel: C₈H₁₅F₃N₂O
Molecular Weight: 212.213 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02081939
CAS-Nummer: 477847-10-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR