2-Chloro-5-fluoro-1H-indole-3-carbaldehyde

Lieferant: Apollo Scientific
Achtung

PC300936-1G PC300936-5G PC300936-250MG
APOSPC300936-1GEA 560 EUR
APOSPC300936-1G APOSPC300936-5G APOSPC300936-250MG
2-Chloro-5-fluoro-1H-indole-3-carbaldehyde
2-Chloro-5-fluoro-1H-indole-3-carbaldehyde
Formel: C₉H₅ClFNO
Molecular Weight: 197,594 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05178217
CAS-Nummer: 535925-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR