2-Fluoro-1-methoxy-4-(methylsulphonyl)benzene 97%

Lieferant: Apollo Scientific
Achtung

PC300944-1G PC300944-250MG
APOSPC300944-1GEA 60.9 EUR
APOSPC300944-1G APOSPC300944-250MG
2-Fluoro-1-methoxy-4-(methylsulphonyl)benzene 97%
2-Fluoro-1-methoxy-4-(methylsulphonyl)benzene
Formel: C₈H₉FO₃S
Molecular Weight: 204,219 g/mol
Schmelzpunkt: 97…98 °C
Lagertemperatur: Kühlschrank
MDL: MFCD21090339
CAS-Nummer: 20951-14-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR