2-Chloro-5-fluoro-1-methyl-1H-indole-3-carbaldehyde

Lieferant: Apollo Scientific
Achtung

PC300966-1G PC300966-5G
APOSPC300966-1GEA 416 EUR
APOSPC300966-1G APOSPC300966-5G
2-Chloro-5-fluoro-1-methyl-1H-indole-3-carbaldehyde
2-Chloro-5-fluoro-1-methyl-1H-indole-3-carbaldehyde
Formel: C₁₀H₇ClFNO
Molecular Weight: 211.62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19310038
CAS-Nummer: 1310141-83-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR