2-[4-(Trifluoromethoxy)phenoxy]nicotinonitrile
Lieferant: Apollo Scientific
PC300976-1G
PC300976-5G
APOSPC300976-1GEA
282
EUR
APOSPC300976-1G
APOSPC300976-5G
2-[4-(Trifluoromethoxy)phenoxy]nicotinonitrile
2-[4-(Trifluoromethoxy)phenoxy]nicotinonitrile
Formel:
C₁₃H₇F₃N₂O₂ Molecular Weight: 280,202 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31380628 CAS-Nummer: 1508040-43-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...