(S)-(-)-2-(Fluorodiphenylmethyl)pyrrolidine 95%

Lieferant: Apollo Scientific
Achtung

PC301001-100MG PC301001-250MG
APOSPC301001-100MGEA 0 EUR
APOSPC301001-100MG APOSPC301001-250MG
(S)-(-)-2-(Fluorodiphenylmethyl)pyrrolidine 95%
(S)-(-)-2-(Fluorodiphenylmethyl)pyrrolidine
Formel: C₁₇H₁₈FN
Molecular Weight: 255,33 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03093547
CAS-Nummer: 274674-23-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR