N-(4-Fluorophenyl)benzamide
Lieferant: Apollo Scientific
PC301026-1G
PC301026-5G
APOSPC301026-1GEA
125
EUR
APOSPC301026-1G
APOSPC301026-5G
N-(4-Fluorophenyl)benzamide
N-(4-Fluorophenyl)benzamide
Formel:
C₁₃H₁₀FNO Molecular Weight: 215,223 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017938 CAS-Nummer: 366-75-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...