3-Chloro-4-fluoro-6-nitrotoluene 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Chloro-2-fluoro-5-methyl-4-nitrobenzene
PC301204-1G
PC301204-5G
PC301204-10G
PC301204-25G
APOSPC301204-1GEA
34.8
EUR
APOSPC301204-1G
APOSPC301204-5G
APOSPC301204-10G
APOSPC301204-25G
3-Chloro-4-fluoro-6-nitrotoluene 97%
3-Chloro-4-fluoro-6-nitrotoluene
Formel:
C₇H₅ClFNO₂ Molecular Weight: 189,572 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21603714 CAS-Nummer: 1352457-29-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...