2-Fluoro-4,6-bis(trifluoromethyl)benzyl alcohol 98%
Lieferant: Apollo Scientific
PC302073-1G
PC302073-5G
APOSPC302073-1GEA
69.6
EUR
APOSPC302073-1G
APOSPC302073-5G
2-Fluoro-4,6-bis(trifluoromethyl)benzyl alcohol 98%
2-Fluoro-4,6-bis(trifluoromethyl)benzyl alcohol
Formel:
C₉H₅F₇O Molecular Weight: 262,124 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258672 CAS-Nummer: 1017778-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...