3-Chloro-2-fluoro-4-(trifluoromethyl)benzyl alcohol 98%

Lieferant: Apollo Scientific

PC302089-1G PC302089-5G
APOSPC302089-1GEA 246 EUR
APOSPC302089-1G APOSPC302089-5G
3-Chloro-2-fluoro-4-(trifluoromethyl)benzyl alcohol 98%
3-Chloro-2-fluoro-4-(trifluoromethyl)benzyl alcohol
Formel: C₈H₅ClF₄O
Molecular Weight: 228.571 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23795328
CAS-Nummer: 1431329-79-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR