4-Chloro-3-fluoro-5-(trifluoromethyl)benzyl alcohol 98%

Lieferant: Apollo Scientific

PC302105-1G PC302105-5G
APOSPC302105-1GEA 174 EUR
APOSPC302105-1G APOSPC302105-5G
4-Chloro-3-fluoro-5-(trifluoromethyl)benzyl alcohol 98%
4-Chloro-3-fluoro-5-(trifluoromethyl)benzyl alcohol
Formel: C₈H₅ClF₄O
Molecular Weight: 228,571 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23795333
CAS-Nummer: 1431329-65-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR