2,6-Difluoro-3-methyl-DL-phenylalanine

Lieferant: Apollo Scientific

PC302123-1G PC302123-5G
APOSPC302123-1GEA 116 EUR
APOSPC302123-1G APOSPC302123-5G
2,6-Difluoro-3-methyl-DL-phenylalanine
2,6-Difluoro-3-methyl-DL-phenylalanine
Formel: C₁₀H₁₁F₂NO₂
Molecular Weight: 215.197 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660183
CAS-Nummer: 1043500-56-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR