2-Chloro-3-(trifluoromethyl)-DL-phenylalanine
Lieferant: Apollo Scientific
PC302124-1G
PC302124-5G
PC302124-10G
APOSPC302124-1GEA
116
EUR
APOSPC302124-1G
APOSPC302124-5G
APOSPC302124-10G
2-Chloro-3-(trifluoromethyl)-DL-phenylalanine
2-Chloro-3-(trifluoromethyl)-DL-phenylalanine
Formel:
C₁₀H₉ClF₃NO₂ Molecular Weight: 267.632 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115993 CAS-Nummer: 1259973-98-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...