2-Chloro-4,5-difluoro-DL-phenylalanine

Lieferant: Apollo Scientific

PC302126-1G PC302126-5G PC302126-10G
APOSPC302126-1GEA 116 EUR
APOSPC302126-1G APOSPC302126-5G APOSPC302126-10G
2-Chloro-4,5-difluoro-DL-phenylalanine
2-Chloro-4,5-difluoro-DL-phenylalanine
Formel: C₉H₈ClF₂NO₂
Molecular Weight: 235,615 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660323
CAS-Nummer: 1259983-39-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR