2-Chloro-5-fluoro-DL-phenylalanine
Lieferant: Apollo Scientific
PC302129-1G
PC302129-5G
APOSPC302129-1GEA
116
EUR
APOSPC302129-1G
APOSPC302129-5G
2-Chloro-5-fluoro-DL-phenylalanine
2-Chloro-5-fluoro-DL-phenylalanine
Formel:
C₉H₉ClFNO₂ Molecular Weight: 217.625 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660212 CAS-Nummer: 1259958-87-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...