2-Fluoro-5-methyl-DL-phenylalanine

Lieferant: Apollo Scientific

PC302135-1G PC302135-5G
APOSPC302135-1GEA 116 EUR
APOSPC302135-1G APOSPC302135-5G
2-Fluoro-5-methyl-DL-phenylalanine
2-Fluoro-5-methyl-DL-phenylalanine
Formel: C₁₀H₁₂FNO₂
Molecular Weight: 197,206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660228
CAS-Nummer: 1043500-50-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR