3-Fluoro-4-methoxy-DL-phenylalanine

Lieferant: Apollo Scientific

PC302142-1G PC302142-5G
APOSPC302142-1GEA 128 EUR
APOSPC302142-1G APOSPC302142-5G
3-Fluoro-4-methoxy-DL-phenylalanine
3-Fluoro-4-methoxy-DL-phenylalanine
Formel: C₁₀H₁₂FNO₃
Molecular Weight: 213,206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05227943
CAS-Nummer: 603105-73-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR