2-Chloro-6-fluoro-3-methyl-DL-phenylalanine
Lieferant: Apollo Scientific
PC302169-1G
PC302169-5G
PC302169-10G
APOSPC302169-1GEA
116
EUR
APOSPC302169-1G
APOSPC302169-5G
APOSPC302169-10G
2-Chloro-6-fluoro-3-methyl-DL-phenylalanine
2-Chloro-6-fluoro-3-methyl-DL-phenylalanine
Formel:
C₁₀H₁₁ClFNO₂ Molecular Weight: 231.651 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660171 CAS-Nummer: 1259983-62-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...