3-Chloro-5-(trifluoromethoxy)phenylacetonitrile 98%

Lieferant: Apollo Scientific

Synonyme: 2-(3-Chloro-5-(trifluoromethoxy)phenyl)acetonitrile

PC302197-1G PC302197-5G
APOSPC302197-1GEA 328 EUR
APOSPC302197-1G APOSPC302197-5G
3-Chloro-5-(trifluoromethoxy)phenylacetonitrile 98%
3-Chloro-5-(trifluoromethoxy)phenylacetonitrile
Formel: C₉H₅ClF₃NO
Molecular Weight: 235,59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660312
CAS-Nummer: 433938-69-3
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR