2-Chloro-6-fluoro-3-methoxyphenylacetonitrile 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC302234-1G
PC302234-5G
PC302234-10G
APOSPC302234-1GEA
136
EUR
APOSPC302234-1G
APOSPC302234-5G
APOSPC302234-10G
2-Chloro-6-fluoro-3-methoxyphenylacetonitrile 97%
2-Chloro-6-fluoro-3-methoxyphenylacetonitrile
Formel:
C₉H₇ClFNO Molecular Weight: 199,609 g/mol Schmelzpunkt: 53…55 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09832267 CAS-Nummer: 1017777-64-8 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...