4-Methoxy-2-(trifluoromethyl)cinnamic acid 97%
Lieferant: Apollo Scientific
PC302342-1G
PC302342-5G
APOSPC302342-1GEA
197
EUR
APOSPC302342-1G
APOSPC302342-5G
4-Methoxy-2-(trifluoromethyl)cinnamic acid 97%
4-Methoxy-2-(trifluoromethyl)cinnamic acid
Formel:
C₁₁H₉F₃O₃ Molecular Weight: 246,183 g/mol Schmelzpunkt: 202…208 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06205654 CAS-Nummer: 773131-66-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...