5-Chloro-2-(trifluoromethoxy)benzoyl chloride 97%

Lieferant: Apollo Scientific

PC302353-1G PC302353-5G
APOSPC302353-1GEA 231 EUR
APOSPC302353-1G APOSPC302353-5G
5-Chloro-2-(trifluoromethoxy)benzoyl chloride 97%
5-Chloro-2-(trifluoromethoxy)benzoyl chloride
Formel: C₈H₃Cl₂F₃O₂
Molecular Weight: 259,009 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11519355
CAS-Nummer: 1092461-17-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR