5-Fluoro-2-methyl-DL-phenylalanine
Lieferant: Apollo Scientific
PC302361-1G
PC302361-5G
APOSPC302361-1GEA
159
EUR
APOSPC302361-1G
APOSPC302361-5G
5-Fluoro-2-methyl-DL-phenylalanine
5-Fluoro-2-methyl-DL-phenylalanine
Formel:
C₁₀H₁₂FNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660210 CAS-Nummer: 1260003-81-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...