6-Chloro-2-fluoro-3-methoxyphenylacetonitrile 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC302369-1G
PC302369-5G
PC302369-10G
APOSPC302369-1GEA
136
EUR
APOSPC302369-1G
APOSPC302369-5G
APOSPC302369-10G
6-Chloro-2-fluoro-3-methoxyphenylacetonitrile 98%
6-Chloro-2-fluoro-3-methoxyphenylacetonitrile
Formel:
C₉H₇ClFNO Molecular Weight: 199.609 g/mol Schmelzpunkt: 85…87 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09832272 CAS-Nummer: 1017777-80-8 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...