2,5-Bis(trifluoromethyl)benzamide 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC302377-5G
PC302377-1G
APOSPC302377-5GEA
132
EUR
APOSPC302377-5G
APOSPC302377-1G
2,5-Bis(trifluoromethyl)benzamide 97%
2,5-Bis(trifluoromethyl)benzamide
Formel:
C₉H₅F₆NO Molecular Weight: 257,133 g/mol Schmelzpunkt: 166…168 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04973747 CAS-Nummer: 53130-46-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...