3-(2,3-Difluoro-4-methoxyphenyl)propionic acid 97%

Lieferant: Apollo Scientific

PC302392-1G PC302392-5G PC302392-10G
APOSPC302392-1GEA 177 EUR
APOSPC302392-1G APOSPC302392-5G APOSPC302392-10G
3-(2,3-Difluoro-4-methoxyphenyl)propionic acid 97%
3-(2,3-Difluoro-4-methoxyphenyl)propionic acid
Formel: C₁₀H₁₀F₂O₃
Molecular Weight: 216.181 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660223
CAS-Nummer: 886499-84-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR