4-Chloro-3-(trifluoromethyl)benzoyl chloride 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC302411-5G
PC302411-1G
APOSPC302411-5GEA
105
EUR
APOSPC302411-5G
APOSPC302411-1G
4-Chloro-3-(trifluoromethyl)benzoyl chloride 97%
4-Chloro-3-(trifluoromethyl)benzoyl chloride
Formel:
C₈H₃Cl₂F₃O Molecular Weight: 243.01 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115833 CAS-Nummer: 1735-55-3 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...