2-Methoxy-5-(pentafluorothio)benzoic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Methoxy-5-(pentafluorosulphanyl)benzoic acid
PC302467-1G
PC302467-5G
PC302467-10G
APOSPC302467-1GEA
348
EUR
APOSPC302467-1G
APOSPC302467-5G
APOSPC302467-10G
2-Methoxy-5-(pentafluorothio)benzoic acid 97%
2-Methoxy-5-(pentafluorothio)benzoic acid
Formel:
C₈H₇F₅O₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD16652486 CAS-Nummer: 1448319-14-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...