3-(2-Fluoro-5-methylphenyl)propionic acid 98%

Lieferant: Apollo Scientific

Synonyme: 3-(2-Fluoro-5-methylphenyl)propanoic acid

PC302477-1G PC302477-5G
APOSPC302477-1GEA 159 EUR
APOSPC302477-1G APOSPC302477-5G
3-(2-Fluoro-5-methylphenyl)propionic acid 98%
3-(2-Fluoro-5-methylphenyl)propionic acid
Formel: C₁₀H₁₁FO₂
Molecular Weight: 182.192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09025395
CAS-Nummer: 881189-60-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR