3,5-Difluoro-4-methoxy-DL-phenylalanine

Lieferant: Apollo Scientific

PC302494-1G PC302494-5G
APOSPC302494-1GEA 174 EUR
APOSPC302494-1G APOSPC302494-5G
3,5-Difluoro-4-methoxy-DL-phenylalanine
3,5-Difluoro-4-methoxy-DL-phenylalanine
Formel: C₁₀H₁₁F₂NO₃
Molecular Weight: 231,196 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832237
CAS-Nummer: 1259990-37-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR