3-Chloro-4-(trifluoromethyl)-DL-phenylalanine
Lieferant: Apollo Scientific
|
Achtung
|
PC302524-1G
PC302524-5G
PC302524-10G
APOSPC302524-1GEA
148
EUR
APOSPC302524-1G
APOSPC302524-5G
APOSPC302524-10G
3-Chloro-4-(trifluoromethyl)-DL-phenylalanine
3-Chloro-4-(trifluoromethyl)-DL-phenylalanine
Formel:
C₁₀H₉ClF₃NO₂ Molecular Weight: 267.632 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11519330 CAS-Nummer: 1259966-92-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...