3-Chloro-5-(trifluoromethyl)-DL-phenylalanine

Lieferant: Apollo Scientific
Achtung

PC302534-1G PC302534-5G
APOSPC302534-1GEA 148 EUR
APOSPC302534-1G APOSPC302534-5G
3-Chloro-5-(trifluoromethyl)-DL-phenylalanine
3-Chloro-5-(trifluoromethyl)-DL-phenylalanine
Formel: C₁₀H₉ClF₃NO₂
Molecular Weight: 267,632 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115857
CAS-Nummer: 1259961-84-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR