4-Chloro-3,5-difluorophenylacetic acid 98%
Lieferant: Apollo Scientific
PC302577-1G
PC302577-5G
PC302577-10G
APOSPC302577-1GEA
224
EUR
APOSPC302577-1G
APOSPC302577-5G
APOSPC302577-10G
4-Chloro-3,5-difluorophenylacetic acid 98%
4-Chloro-3,5-difluorophenylacetic acid
Formel:
C₈H₅ClF₂O₂ Molecular Weight: 206,574 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09925051 CAS-Nummer: 1000566-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...