4-Chloro-3-fluoro-DL-phenylalanine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-3-(4-chloro-3-fluorophenyl)propanoic acid

PC302580-1G PC302580-5G
APOSPC302580-1GEA 145 EUR
APOSPC302580-1G APOSPC302580-5G
4-Chloro-3-fluoro-DL-phenylalanine
4-Chloro-3-fluoro-DL-phenylalanine
Formel: C₉H₉ClFNO₂
Molecular Weight: 217.625 g/mol
Schmelzpunkt: 243…245 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD06660180
CAS-Nummer: 439587-16-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR