5-Chloro-2-fluoro-DL-phenylalanine

Lieferant: Apollo Scientific

PC302616-1G PC302616-5G PC302616-10G
APOSPC302616-1GEA 116 EUR
APOSPC302616-1G APOSPC302616-5G APOSPC302616-10G
5-Chloro-2-fluoro-DL-phenylalanine
5-Chloro-2-fluoro-DL-phenylalanine
Formel: C₉H₉ClFNO₂
Molecular Weight: 217.625 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660179
CAS-Nummer: 1259966-96-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR